missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanethiol, 98%

Catalog No. AAL0707914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0707914 25 g
AAL0707922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0707914 Supplier Thermo Scientific Chemicals Supplier No. L0707914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1639-09-4 | C7H16S | 132.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1639-09-4
Molecular Formula C7H16S
Molecular Weight (g/mol) 132.265
MDL Number MFCD00004911
InChI Key VPIAKHNXCOTPAY-UHFFFAOYSA-N
Synonym 1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan
PubChem CID 15422
IUPAC Name heptane-1-thiol
SMILES CCCCCCCS

Specifications

Melting Point -43°C
Density 0.854
Boiling Point 173°C to 175°C
Flash Point 46°C (114°F)
Refractive Index 1.4515
Quantity 25 g
UN Number UN3336
Beilstein 1731687
Sensitivity Air Sensitive
Formula Weight 132.27
Percent Purity 98%
Chemical Name or Material 1-Heptanethiol
Show More Show Less
Hazard Category H226-H335
Hazard Statement GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
EINECSNumber 216-678-8
RTECSNumber MJ1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only