missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptanesulfonic Acid Sodium Salt, Spectrum™ Chemical
Specifications
Specifications
| Absorbance | 0.05 (0.1 N Solution at 210 nm) |
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 5 g |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.