missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic Acid Sodium Salt, Spectrum™ Chemical

Catalog No. 18608102
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608102 5 g Amber Glass Bottle
18608103 25 g Amber Glass Bottle
18608104 50 g Amber Glass Bottle
18608105 100 g Amber Glass Bottle
18608106 1 kg Amber Glass Bottle
18608107 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608102 Supplier Spectrum Chemical Mfg Cor Supplier No. H10275GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 207300-90-1

Specifications

Absorbance 0.05 (0.1 N Solution at 210 nm)
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.