Learn More
1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade
CAS: 207300-90-1 | C7H15NaO3S·H2O | 220.27 g/mol
Supplier: Thermo Scientific Chemicals 411270250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatographySpecifications
| 1-Heptanesulfonic Acid Sodium Salt Monohydrate | |
| (2.7g in 100mL water) (pH 3), (2.7 g in 100 ml water) (pH 3) at 0nm, 0.06 max. at 254nm, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm | |
| White | |
| Authentic | |
| C7H17NaO4S | |
| 25 g | |
| Solubility in water: Soluble | |
| CCCCCCCS(=O)(=O)[O-].O.[Na+] | |
| 220.259 | |
| 220.27 |
| 207300-90-1 | |
| >300°C | |
| 7.4% to 9% (100°C, 3 hrs) (vacuum) | |
| Glass Bottle | |
| CH3(CH2)6SO3Na·H2O | |
| sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc | |
| XWZCREJRXRKIRQ-UHFFFAOYSA-M | |
| sodium;heptane-1-sulfonate;hydrate | |
| 23687711 | |
| Powder or Flakes |
Chemical Identifiers
| 207300-90-1 | |
| sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc | |
| sodium;heptane-1-sulfonate;hydrate |
| XWZCREJRXRKIRQ-UHFFFAOYSA-M | |
| 23687711 | |
| CCCCCCCS(=O)(=O)[O-].O.[Na+] |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RUO – Research Use Only