missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade

Catalog No. AC411275000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411275000 500 g Glass bottle
AC411270050 5 g Glass bottle
AC411270250 25 g Glass bottle
AC411271000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC411275000 Supplier Thermo Scientific Chemicals Supplier No. 411275000
Get 1 Extra Chemical When You Buy 2
View all promotions

CAS: 207300-90-1 | C7H15NaO3S·H2O | 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography

Chemical Identifiers

CAS 207300-90-1
InChI Key XWZCREJRXRKIRQ-UHFFFAOYSA-M
Synonym sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
PubChem CID 23687711
IUPAC Name sodium;heptane-1-sulfonate;hydrate
SMILES CCCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Absorbance (2.7g in 100mL water) (pH 3), (2.7 g in 100 ml water) (pH 3) at 0nm, 0.06 max. at 254nm, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm
Melting Point >300.0°C
Color White
Loss on Drying 7.4% to 9% (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C7H15NaO3S·H2O
Linear Formula CH3(CH2)6SO3Na·H2O
Quantity 500 g
Solubility Information (10% w/v in water) Clear colorless
Molecular Weight (g/mol) 220.27
Formula Weight 220.27
Physical Form Powder or Flakes
Chemical Name or Material 1-Heptanesulfonic acid sodium salt monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only