missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic Acid, Sodium Salt, Monohydrate, HPLC Grade, 99% min., MilliporeSigma™

Catalog No. p-4382190
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
MHX01012 25 g
MHX01013 100 g
2 options

Catalog No. MHX01012

Supplier: Technidata HX01012

Only null left

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M

Specifications

Color White
Density 391kg/m3
Assay Percent Range 99% min.
Quantity 25 g
Synonym sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
SMILES [Na+].CCCCCCCS([O-])(=O)=O
IUPAC Name sodium heptane-1-sulfonate
Molecular Weight (g/mol) 202.24
PubChem CID 23687711
Formula Weight 22.24
Physical Form Crystalline Solid
Chemical Name or Material 1-Heptanesulfonic Acid, Sodium Salt, Monohydrate, LiChropur™, For Ion Pair Chromatography
Show More Show Less
Recommended Storage Store at +15°C to +25°C