missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic Acid, Sodium Salt, Monohydrate, HPLC Grade, 99% min., MilliporeSigma™

Catalog No. MHX01013
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHX01013 100 g
MHX01012 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MHX01013 Supplier Technidata Supplier No. HX01013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M

Specifications

Melting Point >3°C
Color White
Density 391kg/m3
Assay Percent Range 99% min.
Packaging Plastic Bottle
Quantity 100 g
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;heptane-1-sulfonate
Molecular Weight (g/mol) 202.244
PubChem CID 23672332
Formula Weight 22.24
Percent Purity 99.80%
Grade Not Available
Physical Form Crystalline Solid
Chemical Name or Material 1-Heptanesulfonic Acid, Sodium Salt, Monohydrate, LiChropur™, For Ion Pair Chromatography
Show More Show Less
Recommended Storage Store at +15°C to +25°C