missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Heptanesulfonic acid sodium salt monohydrate, ∽98%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0215123980
Specifications
1-Heptanesulfonic acid sodium salt monohydrate | |
100g | |
∽98% | |
1-heptanesulfonic acid sodium, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptylsodiumsulfonate, sodium 1-heptanesulfonate, sodium heptane sulfonate, sodium heptane-1-sulfonate, sodium heptane-1-sulphonate, sodium heptanesulfonate | |
CCCCCCCS(=O)(=O)[O-].[Na+] | |
202.244 | |
220.27 |
White | |
22767-50-6 | |
C7H15NaO3S | |
REFMEZARFCPESH-UHFFFAOYSA-M | |
sodium;heptane-1-sulfonate | |
23672332 | |
Crystalline Powder |
Chemical Identifiers
22767-50-6 | |
202.244 | |
1-heptanesulfonic acid sodium, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptylsodiumsulfonate, sodium 1-heptanesulfonate, sodium heptane sulfonate, sodium heptane-1-sulfonate, sodium heptane-1-sulphonate, sodium heptanesulfonate | |
sodium;heptane-1-sulfonate |
C7H15NaO3S | |
REFMEZARFCPESH-UHFFFAOYSA-M | |
23672332 | |
CCCCCCCS(=O)(=O)[O-].[Na+] |