missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic Acid Sodium Salt (HPLC), Fisher Chemical™

Catalog No. p-8820001
Change view
Click to view available options
Quantity:
100 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O301325 25 g
O3013100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O301325 Supplier Thermo Fisher Scientific Supplier No. O3013 25
Edge

Improve HPLC separation of basic analytes by ion pairing.

  • High purity
  • Application tested
  • Dissolves easily

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
MDL Number MFCD00007543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
PubChem CID 23672332

Specifications

Alpha Vector Heptanesulfonic Acid Sodium Salt
Absorbance Pass Test
Melting Point 300°C
Color White
Assay Percent Range ≥98 %
Packaging Glass Bottle
Linear Formula CH3(CH2)6SO3Na
Quantity 25 g
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid, sodium salt, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate, 1-heptanesulfonic acid sodium salt
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;heptane-1-sulfonate
Molecular Weight (g/mol) 202.244
Formula Weight 202.24
Percent Purity ≥98%
Grade HPLC
Physical Form Powder or Solid
Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt
Show More Show Less