missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic Acid, Sodium Salt, 98%

Catalog No. p-4509429
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206130050 5 g Glass bottle
AC206130250 25 g Glass bottle
AC206131000 100 g Glass bottle
AC206135000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC206130050 Supplier Thermo Scientific Chemicals Supplier No. 206130050
Only null left

CAS: 22767-50-6 | C7H15NaO3S | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22767-50-6
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300.0°C
Color White
Loss on Drying 9% max. (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H15NaO3S
Linear Formula CH3(CH2)6SO3Na
Quantity 5 g
Beilstein 04, IV, 57
Solubility Information (10% in water) Clear colorless
Molecular Weight (g/mol) 202.25
Formula Weight 202.25
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 1-Heptanesulfonic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 245-210-5

RUO – Research Use Only