missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptadecanol 97.0+%, TCI America™
$150.98 - $490.25
Chemical Identifiers
| CAS | 1454-85-9 |
|---|---|
| Molecular Formula | C17H36O |
| Molecular Weight (g/mol) | 256.474 |
| MDL Number | MFCD00002822 |
| InChI Key | GOQYKNQRPGWPLP-UHFFFAOYSA-N |
| Synonym | 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 |
| PubChem CID | 15076 |
| ChEBI | CHEBI:77470 |
| IUPAC Name | heptadecan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCO |
Chemical Identifiers
| 1454-85-9 | |
| 256.474 | |
| GOQYKNQRPGWPLP-UHFFFAOYSA-N | |
| 15076 | |
| heptadecan-1-ol |
| C17H36O | |
| MFCD00002822 | |
| 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 | |
| CHEBI:77470 | |
| CCCCCCCCCCCCCCCCCO |
Specifications
| 1454-85-9 | |
| White-Yellow | |
| C17H36O | |
| 5 g | |
| GOQYKNQRPGWPLP-UHFFFAOYSA-N | |
| heptadecan-1-ol | |
| 15076 | |
| 256.47 | |
| Crystalline Powder |
| 54°C | |
| 309°C | |
| MFCD00002822 | |
| 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1 | |
| CCCCCCCCCCCCCCCCCO | |
| 256.474 | |
| CHEBI:77470 | |
| ≥97.0% (GC) | |
| 1-Heptadecanol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MI3885000
TSCA : Yes