missing translation for 'onlineSavingsMsg'
Learn More

1-Formylpiperidine 98.0+%, TCI America™

Catalog No. p-7129560
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F023425ML 25 mL
F0234100ML 100 mL
F0234500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F023425ML Supplier TCI America Supplier No. F023425ML
Only null left

Chemical Identifiers

CAS 2591-86-8
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006483
InChI Key FEWLNYSYJNLUOO-UHFFFAOYSA-N
Synonym n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czg
PubChem CID 17429
ChEBI CHEBI:42546
IUPAC Name piperidine-1-carbaldehyde
SMILES C1CCN(CC1)C=O

Specifications

Color Yellow
Boiling Point 222°C
Quantity 25 mL
UN Number 2810
Formula Weight 113.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Formylpiperidine
RTECSNumber TN0380000
TSCA Yes