missing translation for 'onlineSavingsMsg'
Learn More

1-Formylpiperazine 98.0+%, TCI America™

Catalog No. F019425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F019425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F019425G Supplier TCI America Supplier No. F019425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7755-92-2
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005963
InChI Key MSSDTZLYNMFTKN-UHFFFAOYSA-O
Synonym 1-formylpiperazine, 1-piperazinecarboxaldehyde, 1-piperazinecarbaldehyde, formylpiperazine, n-formylpiperazine, 1-formyl piperazine, piperazinecarbaldehyde, l-formylpiperazine, 4-formylpiperazine, n-formyl piperazine
PubChem CID 82191
IUPAC Name 4-formylpiperazin-1-ium
SMILES O=CN1CC[NH2+]CC1

Specifications

Color Yellow
Boiling Point 266°C
Quantity 25 g
Formula Weight 114.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Formylpiperazine