missing translation for 'onlineSavingsMsg'
Learn More

1-Formylpiperazine, tech. 90

Catalog No. AAA1475714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1475714 25 g
AAA1475722 100 g
AAA1475736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1475714 Supplier Thermo Scientific Chemicals Supplier No. A1475714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7755-92-2 | C5H11N2O | 115.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7755-92-2
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005963
InChI Key MSSDTZLYNMFTKN-UHFFFAOYSA-O
Synonym 1-formylpiperazine, 1-piperazinecarboxaldehyde, 1-piperazinecarbaldehyde, formylpiperazine, n-formylpiperazine, 1-formyl piperazine, piperazinecarbaldehyde, l-formylpiperazine, 4-formylpiperazine, n-formyl piperazine
PubChem CID 82191
IUPAC Name piperazine-1-carbaldehyde
SMILES O=CN1CC[NH2+]CC1

Specifications

Density 1.111
Boiling Point 264°C to 266°C
Flash Point 102°C (216°F)
Refractive Index 1.512
Quantity 25 g
Beilstein 110295
Formula Weight 114.15
Percent Purity ≥90%
Chemical Name or Material 1-Formylpiperazine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 231-813-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only