missing translation for 'onlineSavingsMsg'
Learn More

1-Formyl-4-methylpiperazine, 98%

CAS: 7556-55-0 | C6H12N2O | 128.18 g/mol

Supplier:  Thermo Scientific Chemicals L1935814

 View more versions of this product

Catalog No. AAL1935814


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Formyl-4-methylpiperazine
236°C to 238°C
C6H12N2O
MFCD00085930
113020
JQTMGOLZSBTZMS-UHFFFAOYSA-N
4-methylpiperazine-1-carbaldehyde
3269221
128.17
7556-55-0
>110°C (230°F)
1.491
25 g
1-formyl-4-methylpiperazine, n-methyl-n'-formylpiperazine, 4-methylpiperazin-1-yl methanone, 4-methyl-1-piperazinyl-methanone, 4-methyl-piperazin-1-yl-methanone, ods
CN1CCN(CC1)C=O
128.18
CHEBI:44477
98%

Chemical Identifiers

7556-55-0
128.18
JQTMGOLZSBTZMS-UHFFFAOYSA-N
3269221
4-methylpiperazine-1-carbaldehyde
C6H12N2O
MFCD00085930
1-formyl-4-methylpiperazine, n-methyl-n'-formylpiperazine, 4-methylpiperazin-1-yl methanone, 4-methyl-1-piperazinyl-methanone, 4-methyl-piperazin-1-yl-methanone, ods
CHEBI:44477
CN1CCN(CC1)C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only