missing translation for 'onlineSavingsMsg'
Learn More

1-Formyl-4-methylpiperazine, 98%

Catalog No. AAL1935806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1935806 5 g
AAL1935814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1935806 Supplier Thermo Scientific Chemicals Supplier No. L1935806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7556-55-0 | C6H12N2O | 128.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7556-55-0
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00085930
InChI Key JQTMGOLZSBTZMS-UHFFFAOYSA-N
Synonym 1-formyl-4-methylpiperazine, n-methyl-n'-formylpiperazine, 4-methylpiperazin-1-yl methanone, 4-methyl-1-piperazinyl-methanone, 4-methyl-piperazin-1-yl-methanone, ods
PubChem CID 3269221
ChEBI CHEBI:44477
IUPAC Name 4-methylpiperazine-1-carbaldehyde
SMILES CN1CCN(CC1)C=O

Specifications

Boiling Point 236°C to 238°C
Flash Point >110°C (230°F)
Refractive Index 1.491
Quantity 5 g
Beilstein 113020
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 1-Formyl-4-methylpiperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only