missing translation for 'onlineSavingsMsg'
Learn More

(1-Fluorovinyl)methyldiphenylsilane 96.0+%, TCI America™

Catalog No. p-7129725
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05181G 1 g
F05185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05181G Supplier TCI America Supplier No. F05181G

Chemical Identifiers

CAS 257610-49-4
Molecular Formula C15H15FSi
Molecular Weight (g/mol) 242.37
MDL Number MFCD08276379
InChI Key IBMIUWHXFMPYLJ-UHFFFAOYSA-N
Synonym 1-Fluoro-1-(methyldiphenylsilyl)ethylene
PubChem CID 10922738
IUPAC Name (1-fluoroethenyl)(methyl)diphenylsilane
SMILES C[Si](C(F)=C)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 242.37
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (1-Fluorovinyl)methyldiphenylsilane