missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylcyclopentanol, 98+%

Catalog No. AAB2310922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2310922 100 g
AAB2310906 5 g
AAB2310914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2310922 Supplier Thermo Scientific Chemicals Supplier No. B2310922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17356-19-3 | C7H10O | 110.156 g/mol

1-Ethynylcyclopentanol is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylcyclopentanol is used in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 17356-19-3
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00001365
InChI Key LQMDOONLLAJAPZ-UHFFFAOYSA-N
Synonym 1-ethynylcyclopentanol, cyclopentanol, 1-ethynyl, 1-ethynyl-1-cyclopentanol, cyclopentyl ethynyl carbinol, 1-ethynyl-cyclopentan-1-ol, acmc-20abvz, 1-ethinylcyclopentanol, 1-ethynyl-cyclopentanol, cyclopentanol, ethynyl, 1-ethynylcyclopentanol ?
PubChem CID 87074
IUPAC Name 1-ethynylcyclopentan-1-ol
SMILES C#CC1(CCCC1)O

Specifications

Melting Point 23°C to 27°C
Density 0.962
Boiling Point 156°C to 159°C
Flash Point 48°C (118°F)
Odor Irritating
Refractive Index 1.474
Quantity 100 g
UN Number UN1325
Beilstein 1924167
Solubility Information Insoluble in water.
Formula Weight 110.16
Percent Purity ≥98%
Chemical Name or Material 1-Ethynylcyclopentanol
Show More Show Less
Hazard Category H228-H302-H319-H335
Hazard Statement GHS H Statement
H228-H302-H319-H335
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 241-385-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only