missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylcyclohexanol, 99%

Catalog No. p-7045957
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1259122 100 g
AAL1259136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1259122 Supplier Thermo Scientific Chemicals Supplier No. L1259122

CAS: 78-27-3 | C8H12O | 124.183 g/mol

1-Ethynylcyclohexanol is used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylcyclohexanol is used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water (10g/L at 20°C), diethyl ether, alcohols, esters, and ketone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 78-27-3
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00003858
InChI Key QYLFHLNFIHBCPR-UHFFFAOYSA-N
Synonym 1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol
PubChem CID 6525
IUPAC Name 1-ethynylcyclohexan-1-ol
SMILES C#CC1(CCCCC1)O

Specifications

Melting Point 30°C to 33°C
Density 0.973
Boiling Point 180°C
Flash Point 73°C (163°F)
Odor Mild
Refractive Index 1.483
Quantity 100 g
UN Number UN2811
Beilstein 471404
Solubility Information Soluble in water (10g/L at 20°C),diethyl ether,alcohols,esters,and ketone.
Formula Weight 124.18
Percent Purity 99%
Chemical Name or Material 1-Ethynylcyclohexanol
Show More Show Less
Hazard Category H302-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 201-100-9
RTECSNumber GV9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only