missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-pentylbenzene 97.0+%, TCI America™

Catalog No. E05635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05635G 5 g
E056325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05635G Supplier TCI America Supplier No. E05635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79887-10-8
Molecular Formula C13H16
Molecular Weight (g/mol) 172.27
MDL Number MFCD00173884
InChI Key APGNXGIUUTWIRE-UHFFFAOYSA-N
Synonym 4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene
PubChem CID 2775131
IUPAC Name 1-ethynyl-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)C#C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 172.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethynyl-4-pentylbenzene