missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynyl-4-pentylbenzene 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 172.27 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Ethynyl-4-pentylbenzene |
Safety and Handling
| TSCA | No |