missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-hexylbenzene 95.0+%, TCI America™

Catalog No. p-7129071
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05645G 5 g
E056425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05645G Supplier TCI America Supplier No. E05645G

Chemical Identifiers

CAS 79887-11-9
Molecular Formula C14H18
Molecular Weight (g/mol) 186.30
MDL Number MFCD00173883
InChI Key NFPDFDTYANKKIU-UHFFFAOYSA-N
Synonym 1-eth-1-ynyl-4-hexylbenzene, 4-hexyl-1-ethynylbenzene, 4-hexylphenylacetylene, 1-ethynyl-4-hexyl-benzene, 4-ethynyl-1-hexylbenzene, acmc-209pie, 4-n-hexylphenylacetylene, 1-ethynyl-4-n-hexylbenzene
PubChem CID 2775129
IUPAC Name 1-ethynyl-4-hexylbenzene
SMILES CCCCCCC1=CC=C(C=C1)C#C

Specifications

Quantity 5 g
Formula Weight 186.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethynyl-4-hexylbenzene