missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 1-Ethynyl-1-cyclohexanol
Description
1-Ethynyl-1-cyclohexanol
Specifications
Specifications
| CAS | 78-27-3 |
| Melting Point | 30°C to 33°C (lit.) |
| Density | 0.967 g/mL (at 25°C (literature)) |
| Boiling Point | 180°C (lit.) |
| Molecular Formula | C8H12O |
| Linear Formula | HC///CC6H10OH |
| MDL Number | MFCD00003858 |
| Quantity | 5 L |
| Molecular Weight (g/mol) | 124.18 |
| Percent Purity | ≥99% |