missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1-Ethynyl-1-cyclohexanol

Catalog No. E5140620L
Encompass
Change view
Click to view available options
Quantity:
5 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E5140620L 20 L
E514065L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E5140620L Supplier Sigma Aldrich Supplier No. E5140620L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethynyl-1-cyclohexanol

Specifications

CAS 78-27-3
Melting Point 30°C to 33°C (lit.)
Density 0.967 g/mL (at 25°C (literature))
Boiling Point 180°C (lit.)
Molecular Formula C8H12O
Linear Formula HC///CC6H10OH
MDL Number MFCD00003858
Quantity 20 L
Molecular Weight (g/mol) 124.18
Percent Purity ≥99%