missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpyridinium bromide, 99%

Catalog No. AAA1791114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1791114 25 g
AAA1791122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1791114 Supplier Thermo Scientific Chemicals Supplier No. A1791114
Only null left

CAS: 1906-79-2 | C7H10BrN | 188.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1906-79-2
Molecular Formula C7H10BrN
Molecular Weight (g/mol) 188.068
MDL Number MFCD00041999
InChI Key ABFDKXBSQCTIKH-UHFFFAOYSA-M
Synonym 1-ethylpyridinium bromide, 1-ethylpyridin-1-ium bromide, ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide, n-ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide 1:1, pyridineethobromide, ethylpyridiniumbromide, 1-ethyl-pyridiniubromide, n-ethyl pyridinium bromide
PubChem CID 15927
IUPAC Name 1-ethylpyridin-1-ium;bromide
SMILES CC[N+]1=CC=CC=C1.[Br-]

Specifications

Melting Point 117°C to 121°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 188.07
Percent Purity 99%
Chemical Name or Material 1-Ethylpyridinium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-607-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only