missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpiperidine 98.0+%, TCI America™

Catalog No. E0166500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0166500ML 500 mL
E016625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0166500ML Supplier TCI America Supplier No. E0166500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 766-09-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00006507
InChI Key HTLZVHNRZJPSMI-UHFFFAOYSA-N
Synonym n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene
PubChem CID 13007
ChEBI CHEBI:39017
IUPAC Name 1-ethylpiperidine
SMILES CCN1CCCCC1

Specifications

Color Colorless
Boiling Point 131°C
Quantity 500 mL
UN Number 2386
Formula Weight 113.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Ethylpiperidine
RTECSNumber TN0250000
TSCA Yes