missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpiperazine 98.0+%, TCI America™

Supplier:  TCI America E0355100ML

 View more versions of this product

Catalog No. E0355100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Ethylpiperazine
Yellow
C6H14N2
100 mL
n-ethylpiperazine, piperazine, 1-ethyl, 1-ethylpiperazin, 1-ethyl-piperazine, ethylpiperazine, n-ethyl piperazine, unii-6w2bw0v73g, 1-ethyl-piperazin, ethypiperazine, ethyl piperazine
CCN1CCNCC1
114.192
114.19
Liquid
5308-25-8
158°C
MFCD00059912
2924
WGCYRFWNGRMRJA-UHFFFAOYSA-N
1-ethylpiperazine
79196
≥98.0% (GC,T)

Chemical Identifiers

5308-25-8
114.192
WGCYRFWNGRMRJA-UHFFFAOYSA-N
79196
CCN1CCNCC1
C6H14N2
MFCD00059912
n-ethylpiperazine, piperazine, 1-ethyl, 1-ethylpiperazin, 1-ethyl-piperazine, ethylpiperazine, n-ethyl piperazine, unii-6w2bw0v73g, 1-ethyl-piperazin, ethypiperazine, ethyl piperazine
1-ethylpiperazine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions