missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpiperazine 98.0+%, TCI America™

Catalog No. E0355500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0355500ML 500 mL
E035525ML 25 mL
E0355100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0355500ML Supplier TCI America Supplier No. E0355500ML

Chemical Identifiers

CAS 5308-25-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059912
InChI Key WGCYRFWNGRMRJA-UHFFFAOYSA-N
Synonym n-ethylpiperazine, piperazine, 1-ethyl, 1-ethylpiperazin, 1-ethyl-piperazine, ethylpiperazine, n-ethyl piperazine, unii-6w2bw0v73g, 1-ethyl-piperazin, ethypiperazine, ethyl piperazine
PubChem CID 79196
IUPAC Name 1-ethylpiperazine
SMILES CCN1CCNCC1

Specifications

Color Yellow
Boiling Point 158°C
Quantity 500 mL
UN Number 2924
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Ethylpiperazine