missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpiperazine, 98%

Catalog No. p-7046652
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1553922 100 g
AAL1553936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1553922 Supplier Thermo Scientific Chemicals Supplier No. L1553922
Only null left

CAS: 5308-25-8 | C6H14N2 | 114.192 g/mol

1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5308-25-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059912
InChI Key WGCYRFWNGRMRJA-UHFFFAOYSA-N
Synonym n-ethylpiperazine, piperazine, 1-ethyl, 1-ethylpiperazin, 1-ethyl-piperazine, ethylpiperazine, n-ethyl piperazine, unii-6w2bw0v73g, 1-ethyl-piperazin, ethypiperazine, ethyl piperazine
PubChem CID 79196
IUPAC Name 1-ethylpiperazine
SMILES CCN1CCNCC1

Specifications

Density 0.899
Boiling Point 157°C
Flash Point 43°C (109°F)
Odor Amine-like
Refractive Index 1.469
Quantity 100 g
UN Number UN2733
Beilstein 102971
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 1-Ethylpiperazine
Show More Show Less
Hazard Category H226-H302-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H226
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 226-166-8
RTECSNumber TM0815500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only