missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylcyclohexanol 98.0+%, TCI America™

Catalog No. E11365ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11365ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11365ML Supplier TCI America Supplier No. E11365ML

Chemical Identifiers

CAS 1940-18-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00021402
InChI Key BUCJHJXFXUZJHL-UHFFFAOYSA-N
Synonym 1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
PubChem CID 16021
IUPAC Name 1-ethylcyclohexan-1-ol
SMILES CCC1(O)CCCCC1

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 mL
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethylcyclohexanol
EINECSNumber (3)-2301
RTECSNumber GV9040000
TSCA No