missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylpyridinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. e07565g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07565G Supplier TCI America Supplier No. E07565G
Only null left

Chemical Identifiers

CAS 841251-37-4
Molecular Formula C10H12F6N2O4S2
Molecular Weight (g/mol) 402.326
InChI Key UZIAGXMMNLATPP-UHFFFAOYSA-N
PubChem CID 53384303
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-ethyl-3-methylpyridin-1-ium
SMILES CC[N+]1=CC=CC(=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 402.33
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylpyridinium Bis(trifluoromethanesulfonyl)imide
TSCA No
Recommended Storage Refrigerator