missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Tricyanomethanide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -10°C |
| Quantity | 5 g |
| Formula Weight | 201.23 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Ethyl-3-methylimidazolium Tricyanomethanide |
Safety and Handling
| TSCA | No |