missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Tetrafluoroborate 97.0+%, TCI America™

Catalog No. p-7129030
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04965G 5 g
E049625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04965G Supplier TCI America Supplier No. E04965G
Only null left

Chemical Identifiers

CAS 143314-16-3
Molecular Formula C6H11BF4N2
Molecular Weight (g/mol) 197.97
MDL Number MFCD00216668
InChI Key CUNYTRQQXKCRTJ-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium tetrafluoroborate, 1-ethyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate, emimbf4, basionics™ ee 03, acmc-209cq9, dsstox_cid_29074, dsstox_rid_83293, dsstox_gsid_49218, ksc174g8n, 1-methyl-3-ethylimidazolium tetrafluoroborate
PubChem CID 2769348
IUPAC Name 1-ethyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCN1C=C[N+](C)=C1

Specifications

Melting Point 15°C
Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 197.97
Percent Purity ≥97.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Tetrafluoroborate [for Molten Salt]