missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium p-Toluenesulfonate 98.0+%, TCI America™
Supplier: TCI America E065125G
Specifications
| 1-Ethyl-3-methylimidazolium p-Toluenesulfonate | |
| C13H18N2O3S | |
| HXMUPILCYSJMLQ-UHFFFAOYSA-M | |
| 1-ethyl-3-methylimidazol-3-ium;4-methylbenzenesulfonate | |
| 10636633 | |
| ≥98.0% (HPLC) |
| 328090-25-1 | |
| 25 g | |
| CCN1C=C[N+](=C1)C.CC1=CC=C(C=C1)S(=O)(=O)[O-] | |
| 282.358 | |
| 282.36 | |
| Crystalline Powder |
Chemical Identifiers
| 328090-25-1 | |
| 282.358 | |
| 10636633 | |
| CCN1C=C[N+](=C1)C.CC1=CC=C(C=C1)S(=O)(=O)[O-] |
| C13H18N2O3S | |
| HXMUPILCYSJMLQ-UHFFFAOYSA-M | |
| 1-ethyl-3-methylimidazol-3-ium;4-methylbenzenesulfonate |
Safety and Handling
TSCA : No