missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Methyl Sulfate 98.0+%, TCI America™
$70.84 - $208.78
Chemical Identifiers
| CAS | 516474-01-4 |
|---|---|
| Molecular Formula | C7H14N2O4S |
| Molecular Weight (g/mol) | 222.259 |
| MDL Number | MFCD08457697 |
| InChI Key | BXSDLSWVIAITRQ-UHFFFAOYSA-M |
| Synonym | 1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc |
| PubChem CID | 16211409 |
| IUPAC Name | 1-ethyl-3-methylimidazol-3-ium;methyl sulfate |
| SMILES | CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-] |
Chemical Identifiers
| 516474-01-4 | |
| 222.259 | |
| BXSDLSWVIAITRQ-UHFFFAOYSA-M | |
| 16211409 | |
| CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-] |
| C7H14N2O4S | |
| MFCD08457697 | |
| 1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc | |
| 1-ethyl-3-methylimidazol-3-ium;methyl sulfate |
Specifications
| 516474-01-4 | |
| MFCD08457697 | |
| 1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc | |
| CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-] | |
| 222.259 | |
| 222.26 | |
| Liquid |
| C7H14N2O4S | |
| 5 g | |
| BXSDLSWVIAITRQ-UHFFFAOYSA-M | |
| 1-ethyl-3-methylimidazol-3-ium;methyl sulfate | |
| 16211409 | |
| ≥98.0% (HPLC) | |
| 1-Ethyl-3-methylimidazolium Methyl Sulfate |
Safety and Handling
TSCA : No