missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium methyl sulfate, 98%

Catalog No. AAH5948006
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5948006 5 g
AAH5948018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5948006 Supplier Thermo Scientific Chemicals Supplier No. H5948006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 516474-01-4 | C7H14N2O4S | 222.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 516474-01-4
Molecular Formula C7H14N2O4S
Molecular Weight (g/mol) 222.259
MDL Number MFCD08457697
InChI Key BXSDLSWVIAITRQ-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc
PubChem CID 16211409
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;methyl sulfate
SMILES CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-]

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 222.26
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium methyl sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only