missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Iodide 98.0+%, TCI America™
Supplier: TCI America E055625G
Specifications
| 1-Ethyl-3-methylimidazolium Iodide | |
| 79°C | |
| C6H11IN2 | |
| 25 g | |
| IKQCDTXBZKMPBB-UHFFFAOYSA-M | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium iodide | |
| 11075478 | |
| ≥98.0% (T) |
| 35935-34-3 | |
| Yellow | |
| MFCD03701101 | |
| 1-ethyl-3-methylimidazolium iodide, 1-ethyl-3-methylimidazoliumiodide, 1-ethyl-3-methyl-1h-imidazol-3-ium iodide, acmc-209ik2, dsstox_cid_29137, dsstox_rid_83356, dsstox_gsid_49281, ksc495i0n, 1-ethyl-3-methyl imidazolium iodide salt | |
| [I-].CCN1C=C[N+](C)=C1 | |
| 238.07 | |
| 238.07 | |
| Crystalline Powder |
Chemical Identifiers
| 35935-34-3 | |
| 238.07 | |
| IKQCDTXBZKMPBB-UHFFFAOYSA-M | |
| 11075478 | |
| [I-].CCN1C=C[N+](C)=C1 |
| C6H11IN2 | |
| MFCD03701101 | |
| 1-ethyl-3-methylimidazolium iodide, 1-ethyl-3-methylimidazoliumiodide, 1-ethyl-3-methyl-1h-imidazol-3-ium iodide, acmc-209ik2, dsstox_cid_29137, dsstox_rid_83356, dsstox_gsid_49281, ksc495i0n, 1-ethyl-3-methyl imidazolium iodide salt | |
| 1-ethyl-3-methyl-1H-imidazol-3-ium iodide |
Safety and Handling
TSCA : No