missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium iodide, 97%

Catalog No. p-7053232
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5940406 5 g
AAH5940418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5940406 Supplier Thermo Scientific Chemicals Supplier No. H5940406

CAS: 35935-34-3 | C6H11IN2 | 238.07 g/mol

It is used as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as OLED materials.

Solubility
Soluble in water, , dichloromethane, ethanol, acetonitrile.

Notes
Moisture Sensitive. Keep away from heat and humidity. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 35935-34-3
Molecular Formula C6H11IN2
Molecular Weight (g/mol) 238.07
MDL Number MFCD03701101
InChI Key IKQCDTXBZKMPBB-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium iodide, 1-ethyl-3-methylimidazoliumiodide, 1-ethyl-3-methyl-1h-imidazol-3-ium iodide, acmc-209ik2, dsstox_cid_29137, dsstox_rid_83356, dsstox_gsid_49281, ksc495i0n, 1-ethyl-3-methyl imidazolium iodide salt
PubChem CID 11075478
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;iodide
SMILES [I-].CCN1C=C[N+](C)=C1

Specifications

Color White to Yellow
Quantity 5 g
Beilstein 5160333
Sensitivity Moisture sensitive
Solubility Information Soluble in water,,dichloromethane,ethanol,acetonitrile.
Formula Weight 238.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only