Learn More
1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%
CAS: 412009-61-1 | C6H12N2O4S | 208.23 g/mol
$243.62 - $787.33
Chemical Identifiers
| CAS | 412009-61-1 |
|---|---|
| Molecular Formula | C6H12N2O4S |
| Molecular Weight (g/mol) | 208.23 |
| MDL Number | MFCD06798195 |
| InChI Key | HZKDSQCZNUUQIF-UHFFFAOYSA-M |
| Synonym | 1-ethyl-3-methylimidazolium hydrogen sulfate, 1-ethyl-3-methylimidazolium hydrogensulfate, basionics™ ac 25, acmc-209jj9, dsstox_cid_29156, dsstox_rid_83375, dsstox_gsid_49300, 1-ethyl-3-methylimidazoliumhydrogensulfate, 1-methyl-3-ethylimidazolium hydrogensulfate, 1h-imidazolium,1-ethyl-3-methyl-,sulfate 1:1 |
| PubChem CID | 16217215 |
| SMILES | OS([O-])(=O)=O.CCN1C=C[N+](C)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2723218
|
Thermo Scientific Chemicals
H2723218 |
50 g |
Each for $243.62
|
|
|||||
|
AAH2723230
|
Thermo Scientific Chemicals
H2723230 |
250 g |
Each for $787.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 412009-61-1 | |
| 208.23 | |
| HZKDSQCZNUUQIF-UHFFFAOYSA-M | |
| 16217215 |
| C6H12N2O4S | |
| MFCD06798195 | |
| 1-ethyl-3-methylimidazolium hydrogen sulfate, 1-ethyl-3-methylimidazolium hydrogensulfate, basionics™ ac 25, acmc-209jj9, dsstox_cid_29156, dsstox_rid_83375, dsstox_gsid_49300, 1-ethyl-3-methylimidazoliumhydrogensulfate, 1-methyl-3-ethylimidazolium hydrogensulfate, 1h-imidazolium,1-ethyl-3-methyl-,sulfate 1:1 | |
| OS([O-])(=O)=O.CCN1C=C[N+](C)=C1 |
Specifications
| 412009-61-1 | |
| C6H12N2O4S | |
| 50 g | |
| 1-ethyl-3-methylimidazolium hydrogen sulfate, 1-ethyl-3-methylimidazolium hydrogensulfate, basionics™ ac 25, acmc-209jj9, dsstox_cid_29156, dsstox_rid_83375, dsstox_gsid_49300, 1-ethyl-3-methylimidazoliumhydrogensulfate, 1-methyl-3-ethylimidazolium hydrogensulfate, 1h-imidazolium,1-ethyl-3-methyl-,sulfate 1:1 | |
| OS([O-])(=O)=O.CCN1C=C[N+](C)=C1 | |
| 208.23 | |
| 208.24 | |
| 1-Ethyl-3-methylimidazolium hydrogen sulfate |
| 1.367 | |
| MFCD06798195 | |
| UN3265 | |
| HZKDSQCZNUUQIF-UHFFFAOYSA-M | |
| 1-ethyl-3-methylimidazol-3-ium;hydrogen sulfate | |
| 16217215 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only