missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Hexafluorophosphate 98.0+%, TCI America™
$194.44 - $583.33
Chemical Identifiers
| CAS | 155371-19-0 |
|---|---|
| Molecular Formula | C6H11F6N2P |
| Molecular Weight (g/mol) | 256.132 |
| MDL Number | MFCD00216703 |
| InChI Key | DPDAKOVGQUGTHH-UHFFFAOYSA-N |
| Synonym | 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc |
| PubChem CID | 2758873 |
| IUPAC Name | 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate |
| SMILES | CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F |
Chemical Identifiers
| 155371-19-0 | |
| 256.132 | |
| DPDAKOVGQUGTHH-UHFFFAOYSA-N | |
| 2758873 | |
| CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F |
| C6H11F6N2P | |
| MFCD00216703 | |
| 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc | |
| 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate |
Specifications
| 155371-19-0 | |
| White | |
| MFCD00216703 | |
| 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc | |
| CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F | |
| 256.132 | |
| 256.13 | |
| Crystalline Powder |
| 61°C | |
| C6H11F6N2P | |
| 5 g | |
| DPDAKOVGQUGTHH-UHFFFAOYSA-N | |
| 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate | |
| 2758873 | |
| ≥98.0% (T) | |
| 1-Ethyl-3-methylimidazolium Hexafluorophosphate [for Molten Salt] |
Safety and Handling
TSCA : No