missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%
CAS: 155371-19-0 | C6H11F6N2P | 256.132 g/mol
$120.04 - $843.82
Chemical Identifiers
| CAS | 155371-19-0 |
|---|---|
| Molecular Formula | C6H11F6N2P |
| Molecular Weight (g/mol) | 256.132 |
| MDL Number | MFCD00216703 |
| InChI Key | DPDAKOVGQUGTHH-UHFFFAOYSA-N |
| Synonym | 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc |
| PubChem CID | 2758873 |
| IUPAC Name | 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate |
| SMILES | CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1976204
|
Thermo Scientific Chemicals
L1976204 |
2 g |
Each for $120.04
|
|
|||||
|
AAL1976209
|
Thermo Scientific Chemicals
L1976209 |
10 g |
Each for $358.36
|
|
|||||
|
AAL1976218
|
Thermo Scientific Chemicals
L1976218 |
50 g |
Each for $843.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 155371-19-0 | |
| 256.132 | |
| DPDAKOVGQUGTHH-UHFFFAOYSA-N | |
| 2758873 | |
| CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F |
| C6H11F6N2P | |
| MFCD00216703 | |
| 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc | |
| 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate |
Specifications
| 155371-19-0 | |
| C6H11F6N2P | |
| 2 g | |
| 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc | |
| CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F | |
| 256.132 | |
| 256.13 | |
| 1-Ethyl-3-methylimidazolium hexafluorophosphate |
| 58°C to 62°C | |
| MFCD00216703 | |
| Hygroscopic | |
| DPDAKOVGQUGTHH-UHFFFAOYSA-N | |
| 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate | |
| 2758873 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only