missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Ethyl Sulfate 98.0+%, TCI America™

Catalog No. p-7129085
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06505G 5 g
E065025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06505G Supplier TCI America Supplier No. E06505G
Only null left

Chemical Identifiers

CAS 342573-75-5
Molecular Formula C8H16N2O4S
Molecular Weight (g/mol) 236.286
InChI Key VRFOKYHDLYBVAL-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics™ lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate
PubChem CID 12095229
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;ethyl sulfate
SMILES CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 236.29
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Ethyl Sulfate