missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium bromide, 98%, Thermo Scientific™

Chemical Identifiers

CAS 65039-08-9
Molecular Formula C6H11BrN2
Molecular Weight (g/mol) 191.07
MDL Number MFCD03427610
InChI Key GWQYPLXGJIXMMV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
PubChem CID 2734235
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;bromide
SMILES [Br-].CCN1C=C[N+](C)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC356670050
SDS View Documents
Thermo Scientific Chemicals
356670050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC356670250
SDS View Documents
Thermo Scientific Chemicals
356670250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC356671000
SDS View Documents
Thermo Scientific Chemicals
356671000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Ionic liquid
Specifications

Chemical Identifiers

65039-08-9
191.07
GWQYPLXGJIXMMV-UHFFFAOYSA-M
2734235
[Br-].CCN1C=C[N+](C)=C1
C6H11BrN2
MFCD03427610
1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
1-ethyl-3-methylimidazol-3-ium;bromide

Specifications

65039-08-9
100.0
97.5% min. (Argentometry)
C6H11BrN2
5g
GWQYPLXGJIXMMV-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;bromide
2734235
98%
97.5
Authentic
Glass bottle
MFCD03427610
1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
[Br-].CCN1C=C[N+](C)=C1
191.07
191.07
1-Ethyl-3-methylimidazolium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only