Learn More
1-Ethyl-3-methylimidazolium bromide, 98+%
Description
An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.
Solubility
Soluble in water, dichloromethane, ethanol, acetonitrile. Insoluble in ethyl acetate, ether, and alkane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
Specifications
| Melting Point | 70°C to 73°C |
| Quantity | 10 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water,dichloromethane,ethanol,acetonitrile. Insoluble in ethyl acetate,ether,and alkane. |
| Formula Weight | 191.07 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 1-Ethyl-3-methylimidazolium bromide |
Safety and Handling
| Hazard Category | H315-H319-H335-H500 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only