missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium Acetate 95.0+%, TCI America™
Supplier: TCI America E088325G
Specifications
| 1-Ethyl-3-methylimidazolium Acetate | |
| C8H14N2O2 | |
| 25 g | |
| XIYUIMLQTKODPS-UHFFFAOYSA-M | |
| 1-ethyl-3-methylimidazol-3-ium;acetate | |
| 11658353 | |
| ≥95.0% (HPLC,T) |
| 143314-17-4 | |
| MFCD06798186 | |
| 1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc | |
| CCN1C=C[N+](=C1)C.CC(=O)[O-] | |
| 170.212 | |
| 170.21 | |
| Liquid |
Chemical Identifiers
| 143314-17-4 | |
| 170.212 | |
| XIYUIMLQTKODPS-UHFFFAOYSA-M | |
| 11658353 | |
| CCN1C=C[N+](=C1)C.CC(=O)[O-] |
| C8H14N2O2 | |
| MFCD06798186 | |
| 1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc | |
| 1-ethyl-3-methylimidazol-3-ium;acetate |
Safety and Handling
TSCA : Yes