missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium acetate, 97%

Catalog No. AAH5927706
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5927706 5 g
AAH5927718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5927706 Supplier Thermo Scientific Chemicals Supplier No. H5927706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 143314-17-4 | C8H14N2O2 | 170.212 g/mol

It is used as a solvent and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent and as pharmaceutical intermediate.

Solubility
Soluble in water, acetonitrile, acetone, dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 143314-17-4
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.212
MDL Number MFCD06798186
InChI Key XIYUIMLQTKODPS-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc
PubChem CID 11658353
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;acetate
SMILES CCN1C=C[N+](=C1)C.CC(=O)[O-]

Specifications

Melting Point -20°C
Density 1.027 g/mL
Flash Point 164°C (327°F)
Refractive Index n20/D 1.502
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,acetonitrile,acetone,dichloromethane.
Formula Weight 170.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium acetate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only