missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-hydroxypiperidine 98.0+%, TCI America™

Catalog No. E037625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E037625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E037625ML Supplier TCI America Supplier No. E037625ML
Only null left

Chemical Identifiers

CAS 13444-24-1
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006508
InChI Key ZNPSUOAGONLMLK-UHFFFAOYSA-N
Synonym 1-ethyl-3-hydroxypiperidine, 1-ethyl-3-piperidinol, n-ethyl-3-piperidinol, 3-piperidinol, 1-ethyl, n-ethyl-3-hydroxypiperidine, n-ethyl-3-hydroxy piperidine, ethyl-3-hydroxypiperidine, 3-hydroxy-n-ethylpiperidine, 3-hydroxy-1-ethylpiperidine, dactilake
PubChem CID 114524
IUPAC Name 1-ethylpiperidin-3-ol
SMILES CCN1CCCC(C1)O

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 129.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-hydroxypiperidine
EINECSNumber (9)-0217
TSCA No