missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-2-pyrrolidone 98.0+%, TCI America™
$78.93 - $369.64
Chemical Identifiers
| CAS | 2687-91-4 |
|---|---|
| Molecular Formula | C6H11NO |
| Molecular Weight (g/mol) | 113.16 |
| MDL Number | MFCD00003199 |
| InChI Key | ZFPGARUNNKGOBB-UHFFFAOYSA-N |
| Synonym | 1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidinone, n-ethylpyrrolidinone, ethyl pyrrolidone, n-ethyl-, a-pyrrolidone, a-pyrrolidone |
| PubChem CID | 17595 |
| IUPAC Name | 1-ethylpyrrolidin-2-one |
| SMILES | CCN1CCCC1=O |
Chemical Identifiers
| 2687-91-4 | |
| 113.16 | |
| ZFPGARUNNKGOBB-UHFFFAOYSA-N | |
| 17595 | |
| CCN1CCCC1=O |
| C6H11NO | |
| MFCD00003199 | |
| 1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidinone, n-ethylpyrrolidinone, ethyl pyrrolidone, n-ethyl-, a-pyrrolidone, a-pyrrolidone | |
| 1-ethylpyrrolidin-2-one |
Specifications
| 2687-91-4 | |
| 218°C | |
| MFCD00003199 | |
| 1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidinone, n-ethylpyrrolidinone, ethyl pyrrolidone, n-ethyl-, a-pyrrolidone, a-pyrrolidone | |
| CCN1CCCC1=O | |
| 113.16 | |
| 113.16 | |
| Liquid |
| Yellow | |
| C6H11NO | |
| 25 mL | |
| ZFPGARUNNKGOBB-UHFFFAOYSA-N | |
| 1-ethylpyrrolidin-2-one | |
| 17595 | |
| ≥98.0% (GC) | |
| 1-Ethyl-2-pyrrolidone |
Safety and Handling
EINECSNumber : (5)-0113
RTECSNumber : UY5769250
TSCA : Yes