missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2-pyrrolidinone, 98%

CAS: 2687-91-4 | C6H11NO | 113.16 g/mol

$127.09 - $127.09

Chemical Identifiers

CAS 2687-91-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00003199
InChI Key ZFPGARUNNKGOBB-UHFFFAOYSA-N
Synonym 1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, 2-pyrrolidinone, 1-ethyl, ethyl pyrrolidone, ethyl pyrrolidone, n-ethyl-, a-pyrrolidone, a-pyrrolidone
PubChem CID 17595
IUPAC Name 1-ethylpyrrolidin-2-one
SMILES CCN1CCCC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC163930250
SDS View Documents
Thermo Scientific Chemicals
163930250
25 g Glass bottle
Each for $127.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2687-91-4
113.16
ZFPGARUNNKGOBB-UHFFFAOYSA-N
17595
CCN1CCCC1=O
C6H11NO
MFCD00003199
1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, 2-pyrrolidinone, 1-ethyl, ethyl pyrrolidone, ethyl pyrrolidone, n-ethyl-, a-pyrrolidone, a-pyrrolidone
1-ethylpyrrolidin-2-one

Specifications

2687-91-4
100.0
0.9980g/mL
91°C
97.5% min. (GC)
C6H11NO
MFCD00003199
09,220
1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, 2-pyrrolidinone, 1-ethyl, ethyl pyrrolidone, ethyl pyrrolidone, n-ethyl-, a-pyrrolidone, a-pyrrolidone
ZFPGARUNNKGOBB-UHFFFAOYSA-N
1-ethylpyrrolidin-2-one
17595
113.16
Liquid
97.5
-75.0°C
212.5°C
Authentic
Glass bottle
1.4650 to 1.4670
25 g
0.998
Solubility in water: soluble. Other solubilities: soluble in organic solvents
CCN1CCCC1=O
113.16
2.1 mPa.s (20°C)
98%
1-Ethyl-2-pyrrolidinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Causes serious eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 220-250-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only