missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2-pyrrolidinone, 98%

CAS: 2687-91-4 | C6H11NO | 113.16 g/mol

Supplier:  Thermo Scientific Chemicals B2454822

 View more versions of this product

Catalog No. AAB2454822

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Ethyl-2-pyrrolidinone
0.992
76°C (169°F)
1.465
100 g
ZFPGARUNNKGOBB-UHFFFAOYSA-N
1-ethylpyrrolidin-2-one
17595
98%
2687-91-4
96°C to 98°C (20 mmHg)
C6H11NO
MFCD00003199
1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidinone, ethyl pyrrolidone, ethyl pyrrolidone, n-ethyl-, n-ethyl-, a-pyrrolidone
CCN1CCCC1=O
113.16
113.16

Chemical Identifiers

2687-91-4
113.16
ZFPGARUNNKGOBB-UHFFFAOYSA-N
17595
CCN1CCCC1=O
C6H11NO
MFCD00003199
1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, n-ethylpyrrolidinone, ethyl pyrrolidone, ethyl pyrrolidone, n-ethyl-, n-ethyl-, a-pyrrolidone
1-ethylpyrrolidin-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P201-P202-P210-P281-P305+P351+P338-P308+P313-P310-P370+P378q-P501c

H227-H318-H360Df

EINECSNumber : 220-250-6

RTECSNumber : UY5769250

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only